Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Right-To-Know Label Blanks

Blank RTK labels featuring a permanent, pressure-sensitive adhesive backing

Specifications

Adhesive Type Permanent, Pressure-sensitive
Format Label
Material Vinyl
Product Type Right-to-Know Blank Label
For Use With (Application) Used for safety, maintenance and general markings within a facility
View More Specs

Products 4
Catalog Number Mfr. No. Height (English) Description Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Description Width (English) Price Quantity & Availability  
19-807-127
View Documents
Brady™
59254
3.5 in. Diamond with Hazard Classification 5 in. N/A N/A N/A
19-807-126
View Documents
Brady™
59242
3.5 in. Color Bar 5 in. N/A N/A N/A
19-807-128
View Documents
Brady™
59255
2 in. Diamond with Hazard Classification 2 in. N/A N/A N/A
19-807-125
View Documents
Brady™
59241
5 in. Color Bar 7 in. N/A N/A N/A
Description

Description

  • Compatible with laser printers only
  • Available with color bar design or diamonds with hazard classification

Specifications

Specifications

Permanent, Pressure-sensitive
Vinyl
Used for safety, maintenance and general markings within a facility
3 to 5 Years
-40 to 180°F
14.4 lb./in.
Label
Right-to-Know Blank Label
Laser printer
25/Pk.
-40°C to +82°C
0.0045 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.