Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Static Resistant Heavy Pads

Catalog No. p-3801776 Shop All Brady Corporation Products
Click to view available options
Capacity (English):
16 gal./Bale
30 gal./Cs.
9 gal./Bale
Dimensions (L x W):
38.1 x 48.2 cm (15 x 19 in.)
76.2 x 76.2 cm (30 x 30 in.)

For use with oil and petroleum-based fluids

  • Treated to resist static electricity build-up
  • Absorb only oil and not water
  • Single-ply
  • Color: White
  • Meets National Fire Protection Association (NFPA) specifications

Specifications

Color White
Capacity (English) 30 gal./Cs.
Material Polypropylene
Type Static-resistant Heavy Pad
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
Packaging Quantity 100/Cs.
Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.