Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Striped Warning Tapes

Catalog No. p-6567009 Shop All Brady Corporation Products
Click to view available options
Color:
Black, White
Black, Yellow
Green, White
Red, White
Width (English):
1 in.
1.5 in.
2 in.
3 in.

Indicates hazardous areas clearly, marks physical hazards

  • Permanent adhesive stands up to tough indoor conditions
  • Colors are sealed beneath protective layer of abrasion resistant clear vinyl
  • Holds on to almost any clean, dry surface

Specifications

Adhesive Type Rubber-based Pressure-Sensitive
Color Black, Yellow
Length (English) 54 ft.
Length (Metric) 16.4 m
Width (English) 1 in.
Width (Metric) 2.54 cm
Material B-950 Vinyl
Type Striped Warning Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.