Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ ToughStripe™ Anti-Slip Floor Safety Sign
Shop All Brady Corporation Products
Click to view available options
Displayed Text:
BIOHAZARD
Chemical Storage
Ear Protection Area
Emergency Exit
Exit
Eye Protection Area
Eye Wash Station
Fire Alarm
Fire Extinguisher
Fire Hose
First Aid
Hard Hat Area
Safety Glasses Required
Safety Goggles Required
Safety Shower
Watch Your Step
Color:
Black
Black on Orange
Black on White
Black on Yellow
Red
White
Shape:
Rectangular
Round
Description
- Backed with high-performance adhesive
- Features a highly visible, glossy appearance for communicating important safety and warning messages to employees
- Clear anti-skid area
- Designed with sections of anti-skid material that help prevent accidental slips, trips and falls
- Easy to remove and apply
- Made of polyester
Specifications
Specifications
| Certifications/Compliance | ASTM ANSI F1679, ANSI A1264.2-2006, OSHA |
| Finish | Gloss |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Rubber |
| Description | ToughStripe - Pre-printed Floor Signs |
| Diameter (English) | 17 in. |
| Diameter (Metric) | 431.80 mm |
| For Use With (Application) | Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements with glossy appearance and clear anti-skid area for communicating important safety and warning messages to employees |
| Max. Temperature (English) | 129.9°F |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
