Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Message Tapes

Catalog No. p-6567170 Shop All Brady Corporation Products
Click to view available options
Displayed Text:
Blank
Caution

Catalog No. 19-103-768

Supplier: Brady™ 58255

Only null left
Add to Cart
Add to Cart

Used to warn against potential hazards or to caution against unsafe practices

  • Holds on almost any clean, dry surface
  • Made of B-950 vinyl with permanent adhesive
  • Colors sealed beneath protective layer of abrasion resistant clear vinyl
  • Can be used in toughest indoor conditions

Certifications

Meet OSHA specification 29 CFR 1910.145(c)(2) for Yellow and Black Caution Tape.

Specifications

Color Black, Yellow
Width (Metric) 7.62 cm
Length (Metric) 18.2 m
Width (English) 3 in.
Length (English) 60 ft.
Displayed Text Caution
Material Vinyl
Type Warning Message Tape
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.