Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Posts

Catalog No. 19807296 Shop All Brady Corporation Products
Click to view available options
Color:
Black
Yellow
Yellow, Magenta
Packaging Quantity:
1 Ea.
6 Pk.

Catalog No. 19-807-296

Supplier: Brady™ 92116

Made of colorful polystyrene and polyethylene, these are a decorative alternative to metal posts

  • Nonconductive; can be safely used in electrically hazardous areas
  • Non-rusting (unlike metal posts)
  • Hollow base can be filled with sand for more stable application
  • Large C-hook holds chain in place on large posts and can be used open or closed for greater security
  • Nonreflective

Specifications

Color Black
Height (Metric) 104.14 cm
For Use With (Application) Used in electrically hazardous areas
Height (English) 41 in.
Packaging Quantity 1 Ea.
Type Non-conductive Warning Post
Material Polyethylene
Diameter (English) Base 16 in.
Diameter (English) Outer 3 in.
Diameter (Metric) Base 40.64 cm
Diameter (Metric) Outer 7.62 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.