Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyJet J2000 Color Label Printer and Lockout Tagout Materials Kit for LINK360™ Software

Catalog No. 17377049
Click to view available options
Color:
Blue, Gray

Get more out of your LINK360™ Safety Software when you add the power of on-demand printing. The BradyJet J2000 Color Label Printer and Lockout Tagout Materials Kit is ideal for procedure management and facility ID.

  • Custom-curated kit of printer, ink, labels and tags has everything you need to add tag and label printing capabilities to your LINK360™ Safety Software solution
  • J2000 Printer produces photo-quality, full-color labels fast while minimizing your inventory of labeling supplies and material waste
  • Compact mid-volume inkjet printer can print energy source labels and lockout tags from LINK360™ plus the full-color labels you need for effective ID and specific regulations, including ANSI, GHS, ISO, BS5609 and arc flash
  • BradyJet J2000 Polyester Tags are ideal for use in general facility labeling and compliant with OSHA 1910.145 for accident prevention as well as OSHA 1910.147 for lockout tagout
  • Blank Valve Tags and BradyJet J2000 Continuous Vinyl Tape let you easily create and apply energy source tags
  • CMY Ink Cartridge provides high color mix for vibrant, long-lasting colors
  • LINK360™ Safety Software sold separately

Specifications

Product Type Label Printer
Color Blue, Gray
Display Type None
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Inkjet
Length (English) 15.3 in.
Width (English) 10.4 in.
Length (Metric) 388.62 mm
Width (Metric) 264.16 mm
Height (English) 7.1 in.
Height (Metric) 180.34 mm
Dimensions (L x W x H) 15.3 x 10.4 x 7.1 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Full Color
Includes LINK360 Starter Bundle, BradyJet J2000 Color Label Printer with Safety and Facility Software, 3.25 x 5.75 in. Polyester Tags, BradyJet J2000 Full Color Ink Cartridge, 1.125 in. x 100 ft. B30 Series Continuous Indoor Outdoor Vinyl Labels, 10 Blank White Valve Tags (1.5 x 3 in.)
Languages Chinese (Simplified), Chinese (Traditional), Dutch, English, French, German, Hindi, Italian, Japanese, Portuguese, Russian, Spanish, Thai, Vietnamese, Czech
Model J2000
Print Speed 2.5 in./sec.
Resolution 4800 dpi
Software Type Brady Workstation, Brady LINK360, Windows-based Driver for 3rd Party Software Use
Connector Types USB
Operating System Windows 7, Windows 8, Windows 10, Windows 11
Temperature Range (English) 59°F to 90°F
Temperature Range (Metric) 15°C to 32°C
Certifications/Compliance UL, FCC Class A (USA); cUL, IC/ISED (Canada); CE Mark, 2014/35/EU LVD, 2014/30/EU-EMCD, 2011/65/EU, 2015/863/EU RoHS2, 2014/53/EU RED (EU); RCM (ANZ)
For Use With (Application) Hazard Communication Identification, Barcode Labeling, Lockout Tagout, Pipe and Valve Marking
Warranty 1 Year
Power Supply Details AC Adaptor
Voltage 100/240 V
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.