Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyJet™ J4000 Chemical-resistant Die-cut Polypropylene Labels

Catalog No. p-7238963 Shop All Brady Corporation Products
Click to view available options
Height (English):
3.35 in.
5.25 in.
6.1 in.
10 in.
11 in.
Width (English):
2.17 in.
4.13 in.
7 in.
8 in.

Catalog No. 17-001-036

Supplier: Brady™ J402607425J

Only null left
Add to Cart
Add to Cart

This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.

The B-7425J polypropylene material of these labels is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required. An ink cartridge is required to print this material.

  • Matte polypropylene with acrylic adhesive is printable with Inkjet
  • Low-cost, full-color GHS labeling option withstands chemical exposure
  • Service temperature of -112 to 158°F (-80 to 70°C)

Specifications

Coating None
Color White
Finish Matte
Format Die-cut
Material Polypropylene (B-7425J)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification, Component and Equipment Labeling, General Identification
For Use With (Equipment) Brady Label Printers
Labels per Row 1 Label
Quantity 340 Labels
Temperature Range (English) -112°F to 158°F
Thickness (English) 0.005 in.
Width (English) 2.17 in.
Width (Metric) 55 mm
Height (English) 3.35 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.