Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bradykinin RB2/BDKRB2 Antibody (471916), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB47251D
Description
Bradykinin RB2/BDKRB2 Monoclonal antibody specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Bradykinin RB2/BDKRB2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
471916 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
B2 bradykinin receptor, B2R, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BRB2, DKFZp686O088 | |
NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession # P30411 | |
0.1 mL | |
GPCR | |
624 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction