Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bradykinin RB2/BDKRB2 Rabbit anti-Human, Mouse, Clone: JU39-04, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275421
Description
Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
Bradykinin RB2/BDKRB2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunoprecipitation 1:10 - 1:500 | |
B2 bradykinin receptor, B2R, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BRB2, DKFZp686O088 | |
Rabbit | |
80 kDa | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation | |
JU39-04 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
BDKRB2 | |
Recombinant protein within Human Bradykinin RB2/BDKRB2 aa 262-391/ 391. (SwissProt: P30411 Human) | |
Protein A purified | |
GPCR | |
624.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction