Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Bradylink Warning Stanchion - Solid Color - Small
Shop All Brady Corporation Products
Click to view available options
Color:
Black
Description
- Posts are non-conductive so they can be safely used in electrically hazardous areas
- Hollow base can be filled with sand for more stable application
- Bradylink Chains sold separately
- Solid color stanchion
Specifications
Specifications
| Color | Black |
| Dimensions (Dia. x H) | 2.5 x 40 in. |
| Packaging Quantity | 1 Ea. |
| Height (English) | 40 in. |
| Width (Metric) | 408.94 mm |
| Type | Stanchion |
| Diameter (English) Base | 14 in. |
| Material | Polyethylene, Polystyrene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction