Learn More
Invitrogen™ BRAF35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592209
Description
Immunogen sequence: MSHGPKQPGA AAAPAGGKAP GQHGGFVVTV KQERGEGPRA GEKGSHEEEP VKKRGWPKGK KRKKILPNGP KAPVTGYVRF LNERREQIRT RHPDLPFPEI TKMLGAEWSK LQPTEKQRYL DEAEREKQQY MKELRAYQQS EAYKMCTEKI QEKKIKKEDS SSGLMNTLLN GHKGGDCDGF STFDVPIFTE EFLDQNKARE AELRRLRKMN VAFEEQNAVL QRHTQSMSSA RERLEQELAL EERRTLALQQ QLQAVRQALT ASFASLPVPG TGETPTLGTL DFYMARLHGA IERDPAQHEK LIVRIKEILA QVASEHL; Positive Samples: HeLa, Jurkat, SKOV3, Mouse kidney, Mouse uterus; Cellular Location: Chromosome, Nucleus.
Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
Specifications
BRAF35 | |
Polyclonal | |
Unconjugated | |
BRAF | |
AW610687; BRAF25; BRAF35; BRCA2-associated factor 35; fc85b02; FLJ26127; high mobility group 20 B; high mobility group 20B; high mobility group box domain containing; high mobility group protein 20 B; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; HMG20B; Hmgx2; HMGXB2; hypothetical protein LOC553572; PP7706; pp8857; si:dkey-110c1.6; SMARCE1R; SMARCE1-related protein; SOXL; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; LOW QUALITY PROTEIN: SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; Unknown (protein for MGC:134468); wu:fc85b02; zgc:110001 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10362, 109880, 114486, 15353, 362825, 673 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15056, P28028, Q9P0W2, Q9Z104 | |
BRAF, HMG20B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human HMG20B (NP_0063302). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.