Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRAF35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527996 Supplier Invitrogen™ Supplier No. PA527996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (95%), Rat (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRAF35
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene BRAF
Gene Accession No. P15056, Q9P0W2
Gene Alias AW610687; BRAF25; BRAF35; BRCA2-associated factor 35; fc85b02; FLJ26127; high mobility group 20 B; high mobility group 20B; high mobility group box domain containing; high mobility group protein 20 B; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; HMG20B; Hmgx2; HMGXB2; hypothetical protein LOC553572; PP7706; pp8857; si:dkey-110c1.6; SMARCE1R; SMARCE1-related protein; SOXL; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; LOW QUALITY PROTEIN: SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; Unknown (protein for MGC:134468); wu:fc85b02; zgc:110001
Gene Symbols BRAF, HMG20B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BRAF35. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10362, 673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.