Learn More
Stoelting™ Brain Punch Set
SDP
Stoelting™ Brain Punch Set features spring loaded nylon expellers and a convenient handle
$973.50 - $1311.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 2
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
10-000-245
SDP
View Documents
Stoelting™
57401
0.25 to 1.25 mm Set of 5
Each for $1,311.00
Only null left
Add to Cart
10-000-246
SDP
View Documents
Stoelting™
57402
1.5 to 2 mm Set of 3
Each for $973.50
Only null left
Add to Cart
Description
Description
  • For the microdissection of frozen brain sections
  • Facilitates the Palkovits Punch technique of dissecting out specific brain nuclei for neurochemistry.
  • Nuclei are punched from frozen brain sections on glass slides, and expelled into the reagents.
  • Spring-loaded expellers avoid risk of contamination that might be caused by expelling with breath
  • Each punch has an electropolished sharp end for smooth cuts and minimal tissue adhesion
  • Smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
  • Slight, ±5%, size variation due to electropolish etching to sharpen
  • Replacement punches available individually
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.