Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brain1/POU3F3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981809
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1809 Supplier Bioss Supplier No. BS11947R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.

Specifications

Antigen Brain1/POU3F3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POU3F3
Gene Accession No. P20264
Gene Alias 8-Oct; Brain-1; Brain-specific homeobox/POU domain protein 1; BRN1; Brn-1; brn-1 protein; class III POU protein POU-domain; Oct-8; Octamer-binding protein 8; Octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; POU3F3; RHS1; Rhs2; Skin1
Gene Symbols POU3F3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Brain1/POU3F3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5455
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.