Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND accu-jet pro Pipette Controller

Catalog No. p-7146481 Shop All BrandTech Scientific Products
Click to view available options
Color:
Dark Blue
Green
Magenta
Royal Blue

Catalog No. 13-689-200

Supplier: BrandTech™ 26300

The BRAND accu-jet pro allows fast, comfortable filling of serological and volumetric pipettes for cell culture and other applications.

Touch sensitive buttons allow intuitive speed control for fine meniscus control. Additional control for small volume pipettes is provided by a speed limiter thumbwheel.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips 0.1-200mL volumetric and serological pipets
Color Dark Blue
Model dark blue
Type Pipette Controller
Includes acuu-jet pro pipette controller, nickel metal hydride battery, two battery compartment covers, wall support, AC adapter, and two spare 0.2um sterile membrane filters
Length (Metric) 23 cm
Width (Metric) 23 cm
Height (Metric) 6.2 cm
Electrical Requirements CEE plug
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.