Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ Four-Clear Sided Disposable Cuvettes for Fluorescence

Catalog No. 14381603
Click to view available options
Quantity:
100
500

BRAND™ UV-transparent cuvettes are the cost-effective alternative to glass or quartz cuvettes, perfect for numerous applications.

  • BRAND™ premium quality four-sided clear disposable cuvettes are tailored for superior fluorescence applications
  • Made of proprietary UV-plastic
  • Optimal for assays in the 230 to 800 nm range, offering virtually no autofluorescence for an excellent signal-to-noise ratio
  • Equipped with a 10 mm light path, these cuvettes are designed to fit most fluorimeters
TRUSTED_SUSTAINABILITY

Specifications

Sterility Non-sterile
Type Fluorimetry Cuvette
Description BRAND four sided clear cuvettes for fluorimetry. Proprietary UV-plastic, pack of 500
Color Transparent
Disposable Single-use
Capacity (Metric) 2.5 to 4.5 mL
Material Proprietary UV-plastic
Length (Metric) 12.5mm
Includes Cuvette only
Quantity 500
Path Length 10 mm
Wavelength Range 230 to 800 nm
Height (Metric) 45 mm
For Use With (Equipment) flourometers
Width (Metric) 12.5 mm
For Use With (Application) Fluorimetry
Lightpath 10 mm
Shape Rectangle
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.