Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ BRAND™ BRANDplates cellGrade™ 96-well PS Microplates
Click to view available options
Color:
Black
Black, Clear Bottom, Clear Bottom
Clear
White
White, Clear Bottom, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Packaging:
1/Pk., 50/Cs.
Description
- Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
- Easily binds serum components to for indirect adhesion of cells
- Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- BIO-CERT™ CELL CULTURE QUALITY means inserts: free of pyrogens, endotoxins, human DNA, RNase, and DNase
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids

Specifications
Specifications
Barcode | No Barcode |
Certifications/Compliance | ISO 11137, DIN EN ISO 10993 |
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Binding Type | Carboxyl and Hydroxyl Groups |
Color | Clear |
Product Type | 96-well microplate |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction