Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRAND™ BRANDplates immunoGrade™ 96-Well Microplates

Catalog No. p-3794698 Shop All BrandTech Scientific Products
Click to view available options
Color:
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Round, C Bottom

An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays

  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment immunoGrade
Total Volume (Metric) Well 330 μL
Well Shape Round
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.