Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ 96-Well Skirted Microplates, BIO-CERT™ PCR QUALITY, Low Profile
Shop All BrandTech Scientific Products
Click to view available options
Color:
Clear
White
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Suitable for most commercially available thermal cyclers
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
- Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
Specifications
Specifications
| Barcode | No Barcode |
| Binding Property | Untreated |
| Certifications/Compliance | DNA, DNase, RNase-free |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | Bagged |
| Color | White |
| Disposable | Single-use |
| Product Type | qPCR 96-well plate |
| Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction