Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates

Catalog No. p-4830335
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Volume (Metric) Well:
330 μL
350 μL
360 μL
Surface Treatment:
Non-treated

Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.

  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Homogenous assays, screenings and storage
Quantity 100/Cs.
RNase Free Yes
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.