Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Volume (Metric) Well:
330 μL
350 μL
360 μL
Surface Treatment:
Non-treated
Description
- Polystyrene surface
- Medium binding, nonsterile
- Standard plate for most applications
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Strip plates available
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottoms available in F- shape
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
- Suited for homogenous assays, screening, and storage.

Specifications
Specifications
Barcode | Includes Barcode |
Certifications/Compliance | ANSI, SLAS |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Sterility | Non-sterile |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction