Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates

Catalog No. 13882211
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)

Nontreated sterile microplates for diverse applications

  • PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottom available in F- shape
  • With lid
  • Available in clear, white or black
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Bacteriological assays, screenings, storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS
Format Standard
Lid With Lid
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
Volume (Metric) Well 360 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
For Use With (Application) Bacteriological assays, screening and for storage
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.