Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates
Shop All BrandTech Scientific Products
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Description
- PS surface
- Medium binding
- Sterilized via beta-radiation
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottom available in F- shape
- With lid
- Available in clear, white or black
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.

Specifications
Specifications
Barcode | Includes Barcode |
Certifications/Compliance | ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS |
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Binding Type | Medium Binding |
Color | Clear |
Material | Polystyrene |
Volume (Metric) Well | 350 μL |
No. of Wells | 96 |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction