Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ PFA Wide-Mouth Reagent Bottles

Catalog No. p-4380272 Shop All BrandTech Scientific Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL

VITLAB PFA wide-mouth reagent bottles are suitable for trace analysis work.

  • Leach-resistant bottles for long-term storage of standards
  • Wide mouth for easier access to contents
  • PFA screw caps help protect against outside contamination and leakage
TRUSTED_SUSTAINABILITY

Specifications

Capacity 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent
Diameter (English) 2.4 in.
Material PFA
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Shape Sloped Shoulders
Height (Metric) 15 cm
Height (English) 5.9 in.
Closure Material PFA
Diameter (Metric) 6.1 cm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.