Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Screw Caps

Catalog No. p-4380260
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
5000 mL

VITLAB™ polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Good chemical resistance for long-term storage of liquids.
  • Screw caps help protect against outside contamination and leakage.
  • Autoclavable at 121 C, according to DIN EN 285
TRUSTED_SUSTAINABILITY

Specifications

Capacity 100mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Transparent
Diameter (English) 2.04 in.
Material Polypropylene
Quantity 20/Pk.
Sterility Non-sterile
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Mouth Narrow
Height (Metric) 10 cm
Product Type Reagent Bottle
Height (English) 3.93 in.
Closure Material Polypropylene
Diameter (Metric) 5.2 cm
Closure Type Screw Cap
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.