Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ BRAND™ Reagent Reservoirs

Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
40 Bags of 5
Individually Wrapped
Packs of 10
Description
- Heavy-gauge polypropylene
- Suitable for all 8- and 12-channel pipetters
- Autoclavable
- Non-sterile version includes lid
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Color | Transparent |
Capacity (Metric) | 60 mL |
Material | Polypropylene |
Sterility | Sterile |
Type | Reagent Reservoir |
For Use With (Application) | For use with multichannel pipets. Without lid. |
Packaging | 40 Bags of 5 |
Quantity | 200/Pk. |
Autoclavable | Autoclavable |
Channels | 1 |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction