Learn More
BrandTech™ BRAND™ Transferpettor™ Replacement Seals
Catalog No. p-50249 Shop All BrandTech Scientific Products
Click to view available options
Volume (Metric):
100 to 500 μL
1000 to 5000 μL
200 to 1000 μL
2000 to 10,000 μL
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
  • Aids in achieving precision and accuracy
  • For the most demanding pipetting operations
  • Transferpettors operates on the positive displacement principle, with piston in direct contact with the aspirated liquid
  • Provided with DE-M marking
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DE-M
For Use With Transferpette/Transferpette S
Material HDPE
Type Seal Kit
Volume (Metric) 1000 to 5000 μL
For Use With (Equipment) 1000 to 5000 μL Transferpettor
Quantity 10/Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.