Learn More
BrandTech™ VACUUBRAND™ Basic Vacuum System Accessory, Supplemental Modules
Catalog No. 13889120 Shop All BrandTech Scientific Products
Click to view available options
Pump Type:
Supplemental Module I
Supplemental Module II
Modules allow convenient upgrading of existing vacuum system
Supplemental Module I
  • Addition of this module to the PC3001 Basic adds a CVC3000 vacuum controller to provide all of the VARIO control functions
  • With the addition of this module, the system will be able find and follow boiling point curves, have single-point hysteresis-free vacuum, have up to ten user programs, interface with a computer via a bi-directional RS232 serial port, plug-and-play installation of peripheral valves via VACUU·BUS, and more
  • All functions are controlled with a large graphic display and the use of a 'turn-and-push' jogwheel
  • Inlet catchpot is also included to preserve the performance of the pump from vacuum line condensate
Supplemental Module II
  • Addition of this module to the PC3001 Basic adds an exhaust condenser and catchpot to allow the capture of solvent that passes through the pump
  • Condenser catchpot capacity is 500 mL
PC3001 Basic can only be upgraded to full VARIO performance with the purchase of Supplemental Module I. A standard CVC3000 controller will not interface with the pump.
TRUSTED_SUSTAINABILITY
Specifications
Includes Emission condenser
For Use With PC3001
Pump Type Supplemental Module II
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.