Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ VITLAB™ Measuring Scoops
In assorted sizes
Supplier: BrandTech™ 39794
Description
- Choose polypropylene or polyethylene
- Metric volume (mL) marked on handle
Specifications
Measuring Scoop | |
8.45 oz. | |
Yes | |
66.04 in. |
Polypropylene | |
250 mL | |
For measuring samples | |
26 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction