Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ PFA Round Bottom Flasks

Catalog No. 14379200
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL

Corrosion- and leach-resistant flasks excellent for rotary evaporation and distillation of high-purity chemicals

  • Manufactured from PFA (a perfluoroalkoxy polymer) for excellent heat and chemical resistance
  • Smooth surfaces simplifies cleaning and reduces risk of sample carryover in trace analysis applications
  • Maximum operating temperature under vacuum at 50°C (122°F)
  • Translucent surfaces for easy solvent monitoring
  • Ideal for rotary evaporation and distillation of high-purity chemicals
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity (Metric) 250 mL
    Material PFA
    For Use With (Application) For rotary evaporation and distillation of high-purity chemicals
    Includes Ground socket
    Product Type Distilling Flask
    Autoclavable Autoclavable
    Disposable Reusable
    Graduated No
    Mouth Style Joint
    Sterility Non-sterile
    Bottom Shape Round
    Show More Show Less
    WARNING: Reproductive Harm - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.