Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps
Shop All BrandTech Scientific Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
GL 32
GL 45
GL 63
Description
- Wide mouth for easier access to contents
- Screw caps help protect agaist outside contamination and leakage
Specifications
Specifications
| Capacity | 67.62 oz., 2000 mL |
| Capacity (Metric) | 2000 mL |
| Capacity (English) | 67.62 oz. |
| Color | Transparent |
| Diameter (English) | 5.15 in. |
| Material | Polypropylene |
| Quantity | 6/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction