Learn More
Branson Ultrasonics™ Ultrasonic Generator System Accessories
Basket for parts to sit safely into the ultrasonic tank without touching tank bottom.
Supplier:  Branson Ultrasonics™ CPN916013
 View more versions of this product
Catalog No. 15-335-122
Description
Description
  • All 304 stainless steel construction makes baskets compatible with most cleaning processes
  • Type 304 stainless steel is acceptable for use with all alkaline solutions and many acids
  • Basket has Electro polished finish
  • Carryover and dragout of cleaning solution are minimized by open design and construction
Specifications
Specifications
Mesh Basket
7000 Series 22 Gal. Tank
Wire Mesh No. 8
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.