Learn More
Branson Ultrasonics™ Ultrasonic Generator System Accessories
Basket for parts to sit safely into the ultrasonic tank without touching tank bottom.
$859.00
Specifications
Type Mesh Basket
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
15-335-120
View Documents
Branson Ultrasonics™
CPN916023
7000 Series 5.2 Gal. Tank Wire Mesh No. 3
Each for $859.00
Only null left
Add to Cart
15-335-122
View Documents
Branson Ultrasonics™
CPN916013
7000 Series 22 Gal. Tank Wire Mesh No. 8 N/A N/A N/A
Description
Description
  • All 304 stainless steel construction makes baskets compatible with most cleaning processes
  • Type 304 stainless steel is acceptable for use with all alkaline solutions and many acids
  • Basket has Electro polished finish
  • Carryover and dragout of cleaning solution are minimized by open design and construction
Specifications
Specifications
Mesh Basket
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.