Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRAP Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142696IR
Description
BRAP Polyclonal antibody specifically detects BRAP in Human samples. It is validated for Western BlotSpecifications
BRAP | |
Polyclonal | |
Western Blot | |
BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein | |
The immunogen for this product maps to a region between residue 560 and 592 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315). | |
0.1 mL | |
Lipid and Metabolism | |
8315 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction