Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559145 Supplier Invitrogen™ Supplier No. PA559145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLVVIRLELA EHSPVPAGFG FSAAAGEMSD EEIKKTTLAS AVACLEGKSP GEKVAIIHQH LGRREMTDVI IETMKSNPDE LKTTVEERKS SEA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Negatively regulates MAP kinase activation by limiting the formation of Raf/MEK complexes probably by inactivation of the KSR1 scaffold protein. Also acts as a Ras responsive E3 ubiquitin ligase that, on activation of Ras, is modified by auto-polyubiquitination resulting in the release of inhibition of Raf/MEK complex formation. May also act as a cytoplasmic retention protein with a role in regulating nuclear transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRAP
Gene Accession No. Q7Z569, Q99MP8
Gene Alias 3010002G07Rik; BRAP; BRAP2; BRCA1 associated protein; BRCA1-associated protein; galectin-2-binding protein; IMP; Impedes mitogenic signal propagation; renal carcinoma antigen NY-REN-63; RING finger protein 52; RING-type E3 ubiquitin transferase BRAP2; RNF52
Gene Symbols BRAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRAP. Recombinant protein control fragment (Product #RP-97663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 687346, 72399, 8315
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.