Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRAP Polyclonal Antibody
BRAP Polyclonal Antibody
Supplier: Thermo Scientific PA5109454
Description
BRAP Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
BRAP | |
Polyclonal | |
Unconjugated | |
BRAP | |
3010002G07Rik; BRAP; BRAP2; BRCA1 associated protein; BRCA1-associated protein; galectin-2-binding protein; IMP; Impedes mitogenic signal propagation; renal carcinoma antigen NY-REN-63; RING finger protein 52; RING-type E3 ubiquitin transferase BRAP2; RNF52 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
687346, 72399, 8315 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.21 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7Z569, Q99MP8 | |
BRAP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human BRAP (NP_006759.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction