Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brass foil, alloy 260, 1.6mm (0.064 in.) thick, Thermo Scientific™
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Brass foil, Alloy 260 |
| Name Note | 1.6mm (0.064 in.) thick |
| Quantity | 150 x 150 mm |
| Odor | Odorless |
| Form | Foil |
| MDL Number | MFCD00171255 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction