Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brassinazole 93.0+%, TCI America™
SDP

Catalog No. B2829100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2829100MG 100 mg
B282910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2829100MG Supplier TCI America Supplier No. B2829100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 224047-41-0
Molecular Formula C18H18ClN3O
Molecular Weight (g/mol) 327.81
MDL Number MFCD08276319
InChI Key YULDTPKHZNKFEY-UHFFFAOYNA-N
Synonym 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol
PubChem CID 15477807
ChEBI CHEBI:73177
IUPAC Name 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 327.81
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Brassinazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.