Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ brat Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144542 Supplier Invitrogen™ Supplier No. PA5144542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

brat Polyclonal Antibody for Western Blot, ELISA

A NHL-domain family protein that functions a translational repressor to inhibit cell proliferation (PubMed:11274060). Plays a central role in translation repression of hb mRNA by being recruited by nanos (nos) and pum to the Nanos Response Element (NRE), a 16 bp sequence in the hb mRNA 3'-UTR (PubMed:11274060). Probably recruited by other proteins to repress translation of other mRNAs in other tissues (PubMed:11274060). Involved in the regulation of ribosomal RNA synthesis and cell growth (PubMed:11807032). Participates in abdominal segmentation and imaginal disk development (PubMed:11274060). During neuroblast division, segregates asymmetrically and inhibits self-renewal of one of the two daughter cells (PubMed:16564014). Together with the asymmetrically segregating transcription factor prospero ensures that the daughter cell will stop growing, exit the cell cycle, and differentiate into neurons possibly by modulating the function of dm in ganglion mother cells (GMC) (PubMed:11807032, PubMed:16564014). Restricts developmental potential of type II intermediary neuronal progenitor (INP) cells playing a role in proliferation and maturation of the neuroblasts (PubMed:24550111, PubMed:14561773, PubMed:22143802, PubMed:18342578).
TRUSTED_SUSTAINABILITY

Specifications

Antigen brat
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene brat
Gene Accession No. Q8MQJ9
Gene Alias Brain tumor protein brat CG10719
Gene Symbols brat
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) brat protein (294-487aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 35197
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.