Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Antibody (KEN) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen BRCA1
Clone KEN
Concentration 1.5 mg/mL
Dilution Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100599SS
SDP
View Documents
Novus Biologicals
NB100599SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100599
SDP
View Documents
Novus Biologicals
NB100599
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

BRCA1
1.5 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
P38398
672
Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
KEN
Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
Human
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
BRCA1
IgG3 κ
Protein A purified
240 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.