Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1 Antibody (KEN), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100599H
Description
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot.Specifications
BRCA1 | |
Monoclonal | |
HRP | |
P38398 | |
BRCA1 | |
Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
KEN | |
Western Blot | |
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
Mouse | |
240 kDa | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
672 | |
Store at 4C in the dark. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction