Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1 Antibody (RAY), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100598AF594
Description
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
BRCA1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
BRCA1 | |
Human BRCA1 corresponding to residues 1314-1864 [UniProtNo. P38398] | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
RAY | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
Mouse | |
0.1 mL | |
672 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction