Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRCA1 Monoclonal Antibody (KEN), Biotin
GREENER_CHOICE

Catalog No. PIMA516050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516050 Supplier Invitrogen™ Supplier No. MA516050
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: U2OS and MCF-7 whole cell lysates.

BRCA1 (breast and ovarian cancer susceptibility protein 1) is a nuclear phosphoprotein that plays a role in maintaining genomic stability and acts as a tumor suppressor in combination with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as BASC (for BRCA1 associated genome surveillance complex). BRCA1 associates with RNA polymerase II, and through the C terminal domain, also interacts with histone deacetylase complex. The BRCA1 protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in the BRCA1 gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene. Many alternatively spliced transcript variants, some of which are disease-associated mutations, have been described for this gene, but the full-length natures of only some of these variants has been described. A related pseudogene, which is also located on chromosome 17, has been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone KEN
Concentration 0.7 mg/mL
Conjugate Biotin
Formulation PBS with 0.05% sodium azide
Gene BRCA1
Gene Accession No. P38398
Gene Alias brca 1; Brca1; brca-1; BRCA1, DNA repair associated; BRCA1/BRCA2-containing complex, subunit 1; BRCAI; BRCC1; breast and ovarian cancer susceptibility protein 1; breast and ovarian cancer sususceptibility protein 1; breast cancer 1; breast cancer 1 early onset; breast cancer 1, early onset; breast cancer associated 1; breast cancer type 1 susceptibility protein; Breast cancer type 1 susceptibility protein homolog; BROVCA1; BROVCA12; Delta772-3095; early onset breast cancer 1; Fanconi anemia, complementation group S; FANCS; IRIS; OTTHUMP00000212147; OTTHUMP00000212148; OTTHUMP00000212149; OTTHUMP00000212150; OTTHUMP00000212151; OTTHUMP00000212155; PNCA4; PPP1R53; protein phosphatase 1, regulatory subunit 53; PSCP; RING finger protein 53; RING-type E3 ubiquitin transferase BRCA1; RNF53; truncated BRCA1; truncated breast and ovarian cancer susceptibility protein 1; truncated breast and ovarian cancer sususceptibility protein 1; truncated breast cancer type 1 susceptibility protein
Gene Symbols BRCA1
Host Species Mouse
Immunogen Human BRCA1 corresponding to residues 1314-1864.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 672
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.