Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1, p Ser1423 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100226PCP
Description
BRCA1 Polyclonal antibody specifically detects BRCA1 in Human samples. It is validated for Flow CytometrySpecifications
BRCA1 | |
Polyclonal | |
Flow Cytometry | |
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human Breast Cancer Gene 1 (GeneID 672) around serine 1423 according to the numbering given in entry NP_009225.1. | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
672 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction