Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582936 Supplier Invitrogen™ Supplier No. PA582936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHPISLSSSK CHDSVVSMFK IENHNDKTVS EKNNKCQLIL QNNIEMTTGT FVEEITENYK RNTENEDNKY TAASRNSHNL EFDGSDSSKN DTVCIHKDET DLLFTDQHNI CLKLSGQFMK EGNTQIKEDL S Highest antigen sequence indentity to the following orthologs: Mouse - 50%, Rat - 46%.

Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRCA2
Gene Accession No. P51587
Gene Alias brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11
Gene Symbols BRCA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRCA2. Recombinant protein control fragment (Product #RP-89275).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.