Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596128 Supplier Invitrogen™ Supplier No. PA596128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.

Breast Cancer Type 2 susceptibility protein (BRCA2) is a tumor-suppressor protein reported to be involved in double-strand break repair, homologous recombination and S-phase checkpoint activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene BRCA2
Gene Accession No. O35923, P51587, P97929
Gene Alias brca 2; BRCA1/BRCA2-containing complex, subunit 2; BRCA2; brca-2; BRCA2 DNA repair associated; BRCA2, DNA repair associated; BRCC2; breast and ovarian cancer susceptibility gene, early onset; breast and ovarian cancer susceptibility protein 2; breast cancer 2; breast cancer 2 tumor suppressor; breast cancer 2, early onset; breast cancer 2, mutation 1, University of Wisconsin-Madison; breast cancer susceptibility protein 2; breast cancer type 2 susceptibility protein; Breast cancer type 2 susceptibility protein homolog; BROVCA2; FACD; FAD; FAD1; FANCB; FANCD; FANCD1; Fanconi anemia group D1 protein; Fanconi anemia group D1 protein homolog; GLM3; mutant BRCA2; PNCA2; RAB163; RP11-298P3.4; XRCC11
Gene Symbols BRCA2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12190, 360254, 675
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.