Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCAA1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126618AF350
Description
BRCAA1 Polyclonal antibody specifically detects BRCAA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BRCAA1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), AT-rich interactive domain-containing protein 4B, BRCAA1DKFZp313M2420, breast cancer-associated antigen 1, Breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, Histone deacetylase complex subunit SAP180, Rb-binding protein homolog, RBBP1L1RBP1L1BCAA, RBP1-like protein, retinoblastoma binding protein 1-like 1, Retinoblastoma-binding protein 1-like 1, SAP180MGC163290, SIN3A-associated protein 180, Sin3-associated polypeptide p180 | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human AT rich interactive domain 4B using the numbering given in entry NP_057458.4 (GeneID 51742). | |
0.1 mL | |
Cancer, DNA Repair | |
51742 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction