Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCAA1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100682MFV450
Description
BRCAA1 Polyclonal antibody specifically detects BRCAA1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BRCAA1 | |
| Polyclonal | |
| Western Blot | |
| 180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), AT-rich interactive domain-containing protein 4B, BRCAA1DKFZp313M2420, breast cancer-associated antigen 1, Breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, Histone deacetylase complex subunit SAP180, Rb-binding protein homolog, RBBP1L1RBP1L1BCAA, RBP1-like protein, retinoblastoma binding protein 1-like 1, Retinoblastoma-binding protein 1-like 1, SAP180MGC163290, SIN3A-associated protein 180, Sin3-associated polypeptide p180 | |
| Reacts with residues 1046-1055 [SSKKQKRSHK] of the 136 kDa human, mouse and rat BRCAA1 protein. | |
| 0.1 mL | |
| Cancer, DNA Repair | |
| 51742 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction