Learn More
Invitrogen™ BRCAA1 Polyclonal Antibody

Description
PA1-4419 detects BRCAA1 in human samples. PA1-4419 has been successfully used in Western blotting procedures. By Western blot, this antibody detects a ~136 kDa protein representing BRCAA1 in MCF-7 cell lysates. PA1-4419 immunizing peptide corresponds to amino acid residues 1046-1055 of human BRCAA1.
Specifications
Specifications
| Antigen | BRCAA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ARID4B |
| Gene Accession No. | Q4LE39 |
| Gene Alias | 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; ARID4B; AT rich interactive domain 4B (Rbp1 like); AT rich interactive domain 4B (RBP1-like); AT-rich interaction domain 4B; AT-rich interactive domain-containing protein 4B; Bcaa; BRCAA1; breast cancer-associated antigen; breast cancer-associated antigen 1; breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; histone deacetylase complex subunit SAP180; Rb-binding protein homolog; RBBP1L1; RBP1L1; retinoblastoma binding protein 1-like 1; retinoblastoma-binding protein 1-like 1; retinoblastoma-binding protein 1-related protein; Sap180; SIN3A-associated protein 180; Sin3-associated polypeptide p180 |
| Gene Symbols | ARID4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.